Aluminum magnesium (alloy 5056) (95:5 %) (w/w), gauze woven from wire 16 mesh Ø 0.24 mm (0.0095 in)
Catalog # 46714.GT
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:1772
- Pk Info:150×150 mm
Specifications
About this item
:
Storage Temperature:Ambient
Formula:Al:Mg;95:5wt%
MDL Number:MFCD00214039