Order Entry
Ireland
ContactUsLinkComponent
Antimony(III) chloride ≥99.0% (by iodometry) for synthesis, Sigma-Aldrich®
Corrosive
Environment
Antimony(III) chloride ≥99.0% (by iodometry) for synthesis, Sigma-Aldrich®
Catalog # 8.14656.9045
Supplier:  Merck
CAS Number:  
Corrosive
Environment
Antimony(III) chloride ≥99.0% (by iodometry) for synthesis, Sigma-Aldrich®
Catalog # 8.14656.9045
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    45 kg
  • Seq:
    4388

Specifications

Specification Test Results

  • Solubility:
    931 g/l (20 °C) Hydrolysis
  • Melting Point:
    73 °C
  • Molar Mass:
    228.1 g/mol
  • Bulk Density:
    1600 kg/m3
  • Boiling Point:
    233 °C (1013 hPa)
  • Vapor Pressure:
    0.16 hPa (20 °C)
  • Density:
    3.14 g/cm3 (20 °C)
  • pH:
    (H2O) acidic
  • Synonyms:
    Antimonous acid, Antimony trichloride
  • Hill Formula:
    Cl3Sb
  • Chemical Formula:
    SbCl3

Specification Test Results

About this item

 :
 :228.12 g/mol
ADR:8,II
 :M-Clarity™ quality level = MQ200
UN:1733
 :GHS05
 :GHS09
CAS Number:10025-91-9
 :MFCD00134030
Formula:SbCl₃
MDL Number:MFCD00011212