Thiamine hydrochloride ≥98.0% FCC
:67-03-8
:
:C₁₂H₁₈Cl₂N₄OS
:MFCD00012780
:337.27 g/mol
:Ambient
- Assay (C₁₂H₁₇ClN₄Os[-HCl):98.0 - 102.0 %
- Color of Solution:To Pass Test
- pH of a 1 in 100 Solution:2.7 - 3.4
- Lead (Pb):max. 2 mg/kg
- Nitrate (NO₃):To Pass Test
Frequently Bought Together