Order Entry
Ireland
ContactUsLinkComponent
Cyclohexane for gas chromatography Plus, Burdick & Jackson™
Cyclohexane for gas chromatography Plus, Burdick & Jackson™
Catalog # HONC65832-7L
CAS Number:  
Cyclohexane for gas chromatography Plus, Burdick & Jackson™
Catalog # HONC65832-7L
CAS Number:  

Specifications

  • Pack type:
    Stainless steel can, returnable
  • Pk:
    7 L
  • Seq:
    5202

Specifications

About this item

 :
 :84.16 g/mol
Storage Temperature:Ambient
EINECS:203-806-2
UN:1145
ADR:3,II
CAS Number:110-82-7
Formula:C₆H₁₂
MDL Number:MFCD00003814