T2-Toxin-solution 100 μg/mL in acetonitrile, Analytical standard, reference material, Fluka™
Catalog # HONCR1966-2ML
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:2 mL
- Standards:Analytical standard
- CAS No.:21259-20-1
- Description:T2-Toxin-solution 100 μg/mL in acetonitrile
- Storage conditions:-20°C
- MDL No.:MFCD00871255
- UN No.:1648
- Synonym(s):[(1S,2R,4S,7R,9R,10R,11S,12S)-11-acetyloxy-2-(acetyloxymethyl)-10-hydroxy-1,5-dimethylspiro[8-oxatricyclo[7.2.1.02,7]dodec-5-ene-12,2'-oxirane]-4-yl] 3-methylbutanoate
- Formula:C₂₄H₃₄O₉
- Concentration:100 μg/mL in acetonitrile
- Description:T2-Toxin
- Flash point:5 °C
- Physical form:Liquid
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery