Potassium bromide ≥99.9995% (metals basis), TraceSELECT™, Fluka™
Catalog # 90737-100G
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Aluminium (Al):Max. 0.02 ppm
- Arsenic (As):Max. 0.01 ppm
- Barium (Ba):Max. 0.5 ppm
- Calcium (Ca):Max. 0.1 ppm
- Cadmium (Cd):Max. 0.005 ppm
- Cerium (Ce):Max. 0.005 ppm
- Cobalt (Co):Max. 0.005 ppm
- Chromium (Cr):Max. 0.005 ppm
- Copper (Cu):Max. 0.005 ppm
- Iron (Fe):Max. 0.01 ppm
- Mercury (Hg):Max. 5 ppb
- Iridium (Ir):Max. 0.5 ppm
- Lanthanum (La):Max. 0.005 ppm
- Lithium (Li):Max. 0.5 ppm
- Magnesium (Mg):Max. 0.05 ppm
- Manganese (Mn):Max. 0.005 ppm
- Sodium (Na):Max. 5 ppm
- Nickel (Ni):Max. 0.005 ppm
- Lead (Pb):Max. 0.005 ppm
- Palladium (Pd):Max. 0.5 ppm
- Platinum (Pt):Max. 0.5 ppm
- Rhodium (Rh):Max. 0.5 ppm
- Ruthenium (Ru):Max. 0.5 ppm
- Antimony (Sb):Max. 0.01 ppm
- Scandium (Sc):Max. 0.005 ppm
- Selenium (Se):Max. 0.01 ppm
- Samarium (Sm):Max. 0.005 ppm
- Strontium (Sr):Max. 0.05 ppm
- Thallium (Tl):Max. 0.01 ppm
- Vanadium (V):Max. 0.5 ppm
- Yttrium (Y):Max. 0.005 ppm
- Zinc (Zn):Max. 0.01 ppm
- Bromate (BrO₃):Max. 10 ppm
- Chloride (Cl):Max. 500 ppm
- Iodide (I):Max. 200 ppm
- Sulfate (SO₄):Max. 10 ppm
- Total N:Max. 2 ppm
Specification Test Results
About this item
:
Boiling Pt:1,383 °C at 1.013 hPa
Melting Pt:734 °C
Density:2,73 g/cm3 (20 °C)
:119 g/mol
Storage Temperature:Ambient
CAS Number:7758-02-3
Formula:KBr
MDL Number:MFCD00011358
Boiling Pt:1,383 °C (1.013 hPa)