Order Entry
Ireland
ContactUsLinkComponent
Cleanroom wipes, sterile, sealed edge, ValuSeal® IonX™
Cleanroom wipes, sterile, sealed edge, ValuSeal® IonX™
Catalog # BERKVSI.0909.14
Supplier:  BERKSHIRE
Cleanroom wipes, sterile, sealed edge, ValuSeal® IonX™
Catalog # BERKVSI.0909.14
Supplier:  BERKSHIRE
Specifications
  • Packed:
    150 sheets/pack
  • Description:
    ValuSeal® IonX® sealed edge cleanroom laundered knit wipe
  • L×W:
    229×229 mm
  • Pk:
    2.100
Specifications
About this item
Designed for the highest level of contamination control in critical processing applications.
  • Critically low particles, fibres, ions, and extractables
  • High abrasion resistance
  • Lightweight, high absorbency material reduces actual cost in use compared to heavier weight products
  • Laser sealed edge for reduced fibre contamination
ValuSeal® IonX® is a laser sealed edge cleanroom laundered wipe recommended for ISO Class 3 and above environments. Composed of 100% polyester continuous filament yarns, its smooth knit construction and laser sealed edges make it ideal for applications requiring critically low levels of particle and fibre release, along with durability and chemical compatibility.
Certifications: ISO Class 3 – 5. Cleanroom Class 1 – 100. EU Grade B/C/D.