Order Entry
Ireland
ContactUsLinkComponent
Nicardipine Related Compounds, Reference Standard
Nicardipine Related Compounds, Reference Standard
Supplier:  USP
Certificates
Nicardipine Related Compounds, Reference Standard
Supplier:  USP

Specifications

  • CAS No.:
    71784-27-5
  • Synonym(s):
    Bis{2-[benzyl(methyl)amino]ethyl} 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate dihydrochloride
  • Description:
    Nicardipine Related Compound D

Specifications

Frequently Bought Together