Order Entry
Ireland
ContactUsLinkComponent
Dehydro Desloratadine, Pharmaceutical Analytical Impurities
Dehydro Desloratadine, Pharmaceutical Analytical Impurities
Catalog # 1A01110.
Supplier:  USP
CAS Number:  
Dehydro Desloratadine, Pharmaceutical Analytical Impurities
Catalog # 1A01110.
Supplier:  USP
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    25 mg
  • Standards:
    Pharmaceutical Analytical Impurities
  • CAS No.:
    117811-20-8
  • Description:
    Dehydro Desloratadine
  • Synonym(s):
    8-chloro-11-(piperidin-4-ylidene)-11H-benzo[5,6]cyclohepta[1,2-b]pyridine
  • Description:
    Dehydro Desloratadine

Specifications

About this item

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.