Order Entry
Ireland
ContactUsLinkComponent
PMPI (4-Maleimidophenyl isocyanate)
PMPI (4-Maleimidophenyl isocyanate)
Catalog # BC18
Supplier:  G-Biosciences
CAS Number:  
undefined
PMPI (4-Maleimidophenyl isocyanate)
Catalog # BC18
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

Specification Test Results

  • Spacer Arm (Å):
    8.7
  • Reactive Toward:
    Sulfhydryl + Hydroxyl
  • Membrane Permeable:
    Not tested
  • Water Soluble:
    No
  • Cleavable/ Reversible:
    No

Specification Test Results

About this item

 :
 :214.177 g/mol
 :GHS07
 :GHS08
CAS Number:123457-83-0
Formula:C₁₁H₆N₂O₃
MDL Number:MFCD00270900