Sulpho-NHS-LC-Biotin, Premium Grade, Pierce™
PIERPG82075
:
- Pk:100 mg
- Seq:2440
- Product Type:Sulfo-NHS-LC-Biotin
- Quantity:100mg/1 g
- Product Line:Pierce™
- Cell Permeability:Cell-Impermeant
- Chemical Reactivity:Amine
- Reactive Moiety:Active Ester, Succinimidyl Ester, Sulfo-NHS Ester
- Solubility:DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water
- Label or Dye:Biotin
- Label Type:Biotin & Analogs
- Spacer:Mid-Length
:
:556.59 g/mol
:Freezer
:127062-22-0
:C₂₀H₂₉N₄NaO₉S₂
:MFCD00078534