Order Entry
Ireland
ContactUsLinkComponent
Micro spatula, stainless steel
Micro spatula, stainless steel
Catalog # 300-0618
Supplier:  Sampling Systems
undefined
Micro spatula, stainless steel
Catalog # 300-0618
Supplier:  Sampling Systems

Specifications

  • Length:
    175 mm
  • Blade width:
    12 mm
  • Pk:
    1
  • Blade length:
    40 mm

Specifications

About this item

Perfect for transferring small amounts of powder.

  • Ideal for transferring powders, granules and cream
  • Used for surface sampling and dispensing
  • Crevice free

The range of stainless steel microspatulas are perfect for transferring very small amounts of powders, creams and granules and are ideal for use in pharma, food and chemical companies. The crevice free stainless steel construction means the product can be easily cleaned between uses.