PMPI (4-Maleimidophenyl isocyanate), Pierce™
PIER28100
:
- Pk:50 mg
- Seq:2370
- Product Line:Pierce™
- Labeling Method:Chemical Labeling
- Crosslinker Type:Heterobifunctional
- Reactive Moiety:Maleimide, Isocyanate
- Spacer:Short
- Form:Solid
- Quantity:50 mg
- Type:Crosslinker
- Solubility:DMF, DMSO
:
:214.177 g/mol
:121…123°C
:GHS08
:GHS07
:Refrigerator
:123457-83-0
:C₁₁H₆N₂O₃
:MFCD00270900