- Concentration:100 ug/ml each component in acetonitrile
- Pack type:Glass ampoule
- Pk:1 mL
- Description:LC Multiresidue Pesticide Standard (Mix 5)
- UN No.:1993
A standard mixture, typically used for food testing, containing the following components: 100 ug/ml each of Acibenzolar-S-methyl [CAS:135158-54-2] ; Bupirimate [CAS:41483-43-6] ; Buprofezin [CAS:69327-76-0] ; Carboxin [CAS:5234-68-4] ; Clethodim [CAS:99129-21-2] ; Clothianidin [CAS:210880-92-5] ; Cyazofamid [CAS:120116-88-3] ; Ethiprole [CAS:181587-01-9] ; Ethofumesate [CAS:26225-79-6] ; Fenamidone [CAS:161326-34-7] ; Fipronil [CAS:120068-37-3] ; Flubendiamide [CAS:272451-65-7] ; Flufenacet [CAS:142459-58-3] ; Hexythiazox [CAS:78587-05-0] ; Mefenacet [CAS:73250-68-7] ; Mesotrione [CAS:104206-82-8] ; Methoprotryne [CAS:841-06-5] ; Metribuzin [CAS:21087-64-9] ; Prometryn [CAS:7287-19-6] ; Propargite [CAS:2312-35-8] ; Prothioconazole [CAS:178928-70-6] ; Pyridaben [CAS:96489-71-3] ; Simetryn [CAS:1014-70-6] ; Sulfentrazone [CAS:122836-35-5] ; Terbutryn [CAS:886-50-0] ; Thiabendazole [CAS:148-79-8] ; Thiacloprid [CAS:111988-49-9] ; Thiamethoxam [CAS:153719-23-4] ; Thiofanox [CAS:39196-18-4] ; Tricyclazole [CAS:41814-78-2] in Acetonitrile
All mixes are supplied with detailed certificate of analysis, chromatogram and 100% shelf-life. The CoAs provide full traceability of preparation, including the dissolution of the raw materials (with their purity), the preparation of the intermediate solutions and the preparation of the final solutions.