Order Entry
Ireland
ContactUsLinkComponent
 
Sulpho-SIAB (Sulphosuccinimidyl (4-iodoacetyl)aminobenzoate), Pierce™
  PIER22327
 :  
undefined
Sulpho-SIAB (Sulphosuccinimidyl (4-iodoacetyl)aminobenzoate), Pierce™
  PIER22327
 :  

 

  • Pk:
    50 mg
  • Seq:
    2370

 

 

  • Product Line:
    Pierce™
  • Labeling Method:
    Chemical Labeling
  • Crosslinker Type:
    Heterobifunctional
  • Reactive Moiety:
    Haloacetyl, Sulfo-NHS Ester
  • Spacer:
    Mid-Length
  • Form:
    Powder
  • Quantity:
    50 mg
  • Type:
    Crosslinker
  • Solubility:
    Water

 

 

 :
 :504.19 g/mol
 :Refrigerator
 :MFCD00054977
 :C₁₃H₁₀O₈N₂SINa