Order Entry
Ireland
ContactUsLinkComponent
LC-SPDP (Succinimidyl 6-[3-(2-pyridylthio)propionamidohexanoate), Pierce™
LC-SPDP (Succinimidyl 6-[3-(2-pyridylthio)propionamidohexanoate), Pierce™
Catalog # PIER21651
CAS Number:  
undefined
LC-SPDP (Succinimidyl 6-[3-(2-pyridylthio)propionamidohexanoate), Pierce™
Catalog # PIER21651
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :425.522 g/mol
Storage Temperature:Refrigerator
CAS Number:158913-22-5
Formula:C₁₈H₂₃N₃O₅S₂
MDL Number:MFCD00083163