Order Entry
Ireland
ContactUsLinkComponent
 
Anti-CA15-3 Mouse Monoclonal Antibody [clone: DBMM1]
  NOVUNBP2-26672
Anti-CA15-3 Mouse Monoclonal Antibody [clone: DBMM1]
  NOVUNBP2-26672

 

  • Antibody type:
    Primary
  • Antigen symbol:
    CA15-3
  • Clonality:
    Monoclonal
  • Clone:
    DBMM1
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Storage buffer:
    Ascites
  • Storage temperature:
    Store at 4 °C. Do not freeze.
  • Immunogen:
    Human breast cancer cell line ZR-75-1
  • Purification:
    Unpurified
  • Pk:
    0,5 mL

 

 

The CA15-3 Antibody (DBMM1) from Novus Biologicals is a mouse monoclonal antibody to CA15-3. This antibody reacts with human. The CA15-3 Antibody (DBMM1) has been validated for the following applications: Immunohistochemistry-paraffin.

Type: Primary
Antigen: CA15-3
Clonality: Monoclonal
Clone: DBMM1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human