Petri Dishes, Glass, Assistent®
Soda-lime glass, with lid. Especially for use as disposables.
- Good chemical resistance
- Heatable up to +60 °C
- Not suitable for applications related to strong heat or rapid temperature changes
- Ideal for bacterial culture production
- Single compartment
- No vents
Frequently Bought Together