Anti-TNFA Rabbit Monoclonal Antibody [clone: 023]
Supplier: Sino Biological
Certificates
About this item
Anti-TNF-alpha antibody
Type: Primary
Antigen: TNFA
Clonality: Monoclonal
Clone: Clone: 023
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Specifications
- Catalog No:
- SINO50349-R023-1
- SINO50349-R023-50
- Antigen symbol:
- TNFA
- TNFA
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- Clone: 023
- Clone: 023
- Reactivity:
- Mouse
- Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Storage temperature:
- Can be stored at 2℃-8℃. Stable for twelve months at -20℃ to -80℃. Sodium azide is recommended to avoid contamination (final concentration 0.05%-0.1%). It is toxic to cells and should be disposed of properly. Avoid repeated freeze-thaw cycles.
- Can be stored at 2 - 8 ℃. Stable for twelve months at -20 to -80 ℃. Sodium azide is recommended to avoid contamination (final concentration 0.05%-0.1%). It is toxic to cells and should be disposed of properly. Avoid repeated freeze-thaw cycles.
- Shipping temperature:
- ambient temperature
- ambient temperature
Specifications
Frequently Bought Together