Order Entry
Ireland
ContactUsLinkComponent
Anti-IL-22BP Rabbit Monoclonal Antibody [clone: 001]
Anti-IL-22BP Rabbit Monoclonal Antibody [clone: 001]
Catalog # SINO11025-R001-1
Supplier:  Sino Biological
Anti-IL-22BP Rabbit Monoclonal Antibody [clone: 001]
Catalog # SINO11025-R001-1
Supplier:  Sino Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    IL-22BP
  • Clonality:
    Monoclonal
  • Clone:
    Clone: 001
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Seq:
    01
  • Form:
    Liquid
  • Storage temperature:
    Can be stored at 2 - 8 ℃. Stable for twelve months at -20 to -80 ℃. Sodium azide is recommended to avoid contamination (final concentration 0.05%-0.1%). It is toxic to cells and should be disposed of properly. Avoid repeated freeze-thaw cycles.
  • Shipping temperature:
    ambient temperature
  • Size:
    1 ml
  • Pk:
    1 mL

Specifications

About this item

Anti-IL-22BP antibody

Type: Primary
Antigen: IL-22BP
Clonality: Monoclonal
Clone: Clone: 001
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human