Human IL13 protein
SINO10369-HNAC-1
:
- Pk:1 mg
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Human
- Seq:03
- Storage conditions:–20…−80 °C
- Endotoxin content:<1,0 EU per μg of the protein as determined by the LAL method
- Biological activity:Measured in a cell proliferation assay using TF1 human erythroleukemic cells (Kitamura, T. et al. 1989, J. Cell Physiol. 140:323.). The ED50 for this effect is typically 0,5 - 3,0 ng/ml.
- Gene ID:AAK53823.1
- Reconstitution instructions:A hardcopy of COA with reconstitution instruction is sent along with the products. Please refer to it for detailed information.
- Carrier-free:Y
- Protein synonyms:ALRH,IL-13,P600
- Protein/peptide name:IL13
- Purity:>92% as determined by SDS-PAGE
- Molecular weight:The recombinant human IL13 consists of 112 amino acids and migrates as an approximately 12,5 kDa band in SDS-PAGE under reducing conditions as predicted.
- Sequence:Met1-Asn132
- Formulation:Powder
- Shipping temperature:Ambient
A DNA sequence encoding the human IL13 (AAK53823.1) (Met 1-Asn 132) was expressed and purified.