Human IL12B / P40 protein (his tag)
Supplier: Sino Biological
Certificates
About this item
A DNA sequence encoding the p40 subunit of human IL12, termed as IL12B (NP_002178.2) (Met 1-Ser 328) was fused with a polyhistidine tag at the C-terminus.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:His Tag
- Storage conditions:−20...−80 °C
- Endotoxin content:<1,0 EU per μg of the protein as determined by the LAL method
- Biological activity:Measured by its ability to bind biotinylated recombinant human IL12RB1 in a functional ELISA.
- Gene ID:NP_002178.2
- Reconstitution instructions:A hardcopy of COA with reconstitution instruction is sent along with the products. Please refer to it for detailed information.
- Carrier-free:Y
- Animal-free:Y
- Protein synonyms:CLMF,CLMF2,IL-12B,IL12 p40,IMD28,IMD29,NKSF,NKSF2
- Protein/peptide name:IL12B
- Purity:>95% as determined by SDS-PAGE
- Molecular weight:The recombinant human IL12B consists of 317 amino acids and has a predicted molecular mass of 36,2 kDa. As a result of glycosylation, the apparent molecular mass of rhIL12B is approximately 40-50 kDa in SDS-PAGE under reducing conditions.
- Sequence:Met1-Ser328
- Formulation:Powder
- Shipping temperature:Ambient
Specifications
Frequently Bought Together