Order Entry
Ireland
ContactUsLinkComponent
 
Anti-cGAS mouse monoclonal antibody [clone: 5G10]
  CAYM23853-300
Anti-cGAS mouse monoclonal antibody [clone: 5G10]
  CAYM23853-300

 

  • Antibody type:
    Primary
  • Antigen name:
    Cyclic GMP-AMP synthase
  • Antigen symbol:
    cGAS
  • Clonality:
    Monoclonal
  • Clone:
    5G10
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • UniProtKB:
    Uniprot # Q8N884
  • Storage buffer:
    PBS, pH 7,2, with 0,02% sodium azide
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    −20 °C
  • Immunogen:
    Full length human recombinant protein
  • Purification:
    Protein G-purified IgG
  • Size:
    300 µg
  • Pk:
    300 µG

 

 

Cyclic GMP-AMP (cGAMP) synthase (cGAS; Item No. 22810) is a nucleotidyltransferase located in the cytosol that acts as a cytosolic DNA sensor to detect foreign DNA from microbial pathogens as part of the innate immune response. Upon binding to cytosolic DNA, cGAS produces the cyclic dinucleotide second messenger cGAMP, which activates stimulator of interferon genes (STING), leading to activation of the type I interferon (IFN) pathway. In vitro, fibroblasts, macrophages, and dendritic cells isolated from cGAS knockout (cGAS-/-) mice do not produce type I IFNs following DNA transfection or DNA virus infection.{39163} Similarly, cells containing a frame-shift mutation in the cGAS locus fail to mount an immune response to HIV and other retroviruses.{39164} In vivo, cGAS-/- mice infected with herpes simplex virus 1 (HSV-1) have lower levels of IFN-α and IFN-β, shorter survival times, and higher post-mortem levels of HSV-1 in the brain. Cayman’s cGAS Monoclonal Antibody (Clone 5G10) recognizes the full length human cGAS protein at ~59 kDa.

For immunochemical detection of cGAS

Type: Primary
Antigen: cGAS
Clonality: Monoclonal
Clone: 5G10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2A
Reactivity: Human