Order Entry
Ireland
ContactUsLinkComponent
p38 overexpression lysate (Adult Normal)
p38 overexpression lysate (Adult Normal)
Catalog # NOVUNBL1-12873
Supplier:  Novus Biologicals
CAS Number:  
p38 overexpression lysate (Adult Normal)
Catalog # NOVUNBL1-12873
Supplier:  Novus Biologicals
CAS Number:  

Specifications

  • Cell Format:
    Cryopreserved
  • Pk:
    0,1 mg
  • Tested applications:
    WB

Specifications

About this item

p38 Overexpression Lysate (Adult Normal)

Tested Applications: Western Blot.