Order Entry
Ireland
ContactUsLinkComponent
Mouse IgG2b Isotype Control [clone: MPC-11] (PE (Phycoerythrin)-Cy7®)
Mouse IgG2b Isotype Control [clone: MPC-11] (PE (Phycoerythrin)-Cy7®)
Catalog # 60-4732-U100
Supplier:  Tonbo Biosciences
undefined
Mouse IgG2b Isotype Control [clone: MPC-11] (PE (Phycoerythrin)-Cy7®)
Catalog # 60-4732-U100
Supplier:  Tonbo Biosciences

Specifications

  • Antibody type:
    Control
  • Antigen name:
    Mitochondrial pyruvate carrier 11
  • Clone:
    MPC-11
  • Conjugation:
    PE (Phycoerythrin)-Cy7®
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
  • Environmentally Preferable:
  • Storage temperature:
    Store undiluted at 2...8 °C, protect from light, do not freeze.
  • Tested applications:
    Flow Cytometry
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

The MPC-11 immunoglobulin is useful as an isotype-matched control. The MPC-11 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG2b antibodies.