Membrane protein pituitary tissue lysate (adult normal)
Catalog # NOVUNB820-60579
Supplier: Novus Biologicals
CAS Number:
Specifications
- Cell Format:Cryopreserved
- Pk:0,1 mg
- Tested applications:WB, ImmunoPrecipitation
Specifications
About this item
Membrane protein pituitary tissue lysate (adult normal).
Tested applications: Western blot, immunoprecipitation.
Tested reactivity: Human