Order Entry
Ireland
ContactUsLinkComponent
Zinc, powder, AnalaR NORMAPUR® 45 µm for analysis
Zinc, powder, AnalaR NORMAPUR® 45 µm for analysis
Supplier:  VWR Chemicals
Certificates
undefined
Zinc, powder, AnalaR NORMAPUR® 45 µm for analysis
Supplier:  VWR Chemicals
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay:
    min. 98%
  • Appearance:
    grayish, heavy powder
  • Arsenic (As):
    max. 0,0001%
  • Cadmium (Cd):
    max. 0,05%
  • Copper (Cu):
    max. 0.005%
  • Iron (Fe):
    max. 0,005%
  • Lead (Pb):
    max. 0,005%
  • Tin (Sn):
    max. 0,001%

Specification Test Results

Related Information

Frequently Bought Together