Order Entry
Ireland
ContactUsLinkComponent
VWR®, Sterile Alliance Sleeves
VWR®, Sterile Alliance Sleeves
Catalog # 76491-706
Supplier:  VWR Collection
VWR®, Sterile Alliance Sleeves
Catalog # 76491-706
Supplier:  VWR Collection
Specifications
  • Sterile:
    +
  • Colour:
    White
  • Description:
    Without thumbloop
  • Length:
    24"
  • Seq:
    01
  • Pk:
    200
Specifications
About this item
VWR® Made in USA sterile sleeves are an ideal option for single-use needs. Made with the specially formulated alliance breathable microporous fabric.
  • Individually bagged
  • Available with thumb loop configuration
  • ETO sterilised
  • Made with alliance breathable microporous fabric
  • Made in USA
A key part of the VWR® Made in USA PPE line, these sterile cleanroom sleeves provide a comfortable fit while meeting sterility and filtration requirements of many controlled environments.
The Alliance fabric is designed to lower particulate levels by providing extremely smooth outer fabric surface that lowers particle cling and build-up in cleanroom applications. Alliance products have a unique antistatic system that assures static dissipation in sensitive cleanroom applications, while providing a liquid-proof barrier that ensures protection from hazardous substances used in biotech and pharmaceutical cleanrooms.
Packaging: Bagged 1 pair per sterile bag, 100 pairs per case. Packed in poly-lined bag in brown corrugated box.