Silica gel, yellow/blue, fractured, GPR RECTAPUR® 2 - 6 mm, with indicator
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7631-86-9
:112926-00-8
Formula:SiO₂
MDL Number:MFCD00011232
:60.08 g/mol
Boiling Pt:2230 °C (1013 hPa)
Melting Pt:> 1600 °C
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Adsorption capacity (H₂O; 25 °C; 40% RH):≥20,0%
- Residual humidity (150 °C):≤3,0%
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC89051.292 spec_VWRC89051.360
Product specification
VWR® for Drying and Filtration
Product Literature
Frequently Bought Together