Order Entry
Ireland
ContactUsLinkComponent
Bürkle Cylindrical Barbed Y-Connectors, PVDF
Bürkle Cylindrical Barbed Y-Connectors, PVDF
Certificates
undefined
Bürkle Cylindrical Barbed Y-Connectors, PVDF
Specifications
  • Chemical resistance:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Max. temperature:
  • 160 °C (320 °F)
  • 160 °C (320 °F)
  • 160 °C (320 °F)
  • 160 °C (320 °F)
  • 160 °C (320 °F)
  • No. of ports:
  • 3
  • 3
  • 3
  • 3
  • 3
  • Sterilisation:
  • Autoclave
  • Autoclave
  • Autoclave
  • Autoclave
  • Autoclave
  • Warranty:
  • 15 Months
  • 15 Months
  • 15 Months
  • 15 Months
  • 15 Months