Order Entry
Ireland
ContactUsLinkComponent
Parker Hannifin Domnick Hunter Air Purification Systems
Parker Hannifin Domnick Hunter Air Purification Systems
Certificates
undefined
Parker Hannifin Domnick Hunter Air Purification Systems
Specifications
  • Max. pressure:
  • 20 bar (290 psi)
  • 16 bar (232 psi)
  • 16 bar (232 psi)
  • 16 bar (232 psi)
  • 16 bar (232 psi)
  • 16 bar (232 psi)
  • Residual oil content:
  • 0,003 ppm at 70 °F (0,003 mg/m³ at 21 °C)
  • 0,5 ppm at 70 °F (0,6 mg/m³ at 21 °C)
  • 0,5 ppm at 70 °F (0,6 mg/m³ at 21 °C)
  • 0,1 ppm at 70 °F (0,01 mg/m³ at 21 °C)
  • 0,1 ppm at 70 °F (0,01 mg/m³ at 21 °C)
  • 0,1 ppm at 70 °F (0,01 mg/m³ at 21 °C)
  • Ø:
  • 9,65 cm (3,8")
  • 9,65 cm (3,8")
  • 12,95 cm (5,1")
  • 7,62 cm (3")
  • 9,65 cm (3,8")
  • 9,65 cm (3,8")
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year