Order Entry
Ireland
ContactUsLinkComponent
Serological pipet, Falcon®
Serological pipet, Falcon®
Catalog # BDAA357551_COS
Supplier:  Corning
Serological pipet, Falcon®
Catalog # BDAA357551_COS
Supplier:  Corning

Specifications

  • Sterile:
    +
  • Capacity:
    10 ml
  • Description:
    Serological pipet, polystyrene, 0,1 ml increments, disposable
  • Pk:
    200

Specifications

About this item

For tissue culture usage or any non-toxic solution that requires aliquoted or aspirating/dispensing in a sterile environment. Nonpyrogenic pipette with 3 ml overfill capacity.

Packaging: Individually packed in a paper/plastic thermoformed package.