Order Entry
Ireland
ContactUsLinkComponent
 
Tetracycline hydrochloride, powder
Catalog # MDTC61-242-RG
Supplier:  Corning
CAS Number:  
undefined
Tetracycline hydrochloride, powder
Catalog # MDTC61-242-RG
Supplier:  Corning
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :480.9 g/mol
Storage Temperature:Ambient
CAS Number:64-75-5
Formula:C₂₂H₂₅ClN₂O₈
MDL Number:MFCD00078142