Order Entry
Ireland
Orders LinkContactUsLinkComponent
VWR®, Storage Boxes for Sample Vials and Bottles, PP
VWR®, Storage Boxes for Sample Vials and Bottles, PP
Supplier:   VWR Collection
VWR®, Storage Boxes for Sample Vials and Bottles, PP
Supplier:  VWR Collection
Certificates
VWR®, Storage Boxes for Sample Vials and Bottles, PP
VWR®, Storage Boxes for Sample Vials and Bottles, PP
15 Options Available Below
Product Details & Documents
A broad variety of PP storage boxes to securely hold a wide range of vial sizes and bottles. Perfect for space-saving storage in the laboratory or in fridges.
  • Transparent lid
  • Chemically resistant
  • Stackable and robust
  • Temperature resistant from –80 to 100 °C
  • Easy sample identification (alphanumeric coding) for boxes with 81 cavities
Transperent lids prevents condensation on the closures and thus avoid contamination in the refrigerator.
Product Family Options
81 Cavities with Alphanumeric Coding
Product Information
  • Colour
    W×D×H
    Availability
    Price
  • 548-1443A
    Box with 81 cavities for 1,5 ml vials
    Box with 81 cavities for 1,5 ml vials
    Blue
    130×130×45 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 35,40 €
    35,40 €
    1 * 1 items
       35,40 €
  • 548-1444A
    Box with 81 cavities for 1,5 ml vials
    Box with 81 cavities for 1,5 ml vials
    Orange
    130×130×45 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 35,40 €
    35,40 €
    1 * 1 items
       35,40 €
  • 548-1445A
    Box with 81 cavities for 1,5 ml vials
    Box with 81 cavities for 1,5 ml vials
    Pink
    130×130×45 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 35,40 €
    35,40 €
    1 * 1 items
       35,40 €
  • 548-1446A
    Box with 81 cavities for 1,5 ml vials
    Box with 81 cavities for 1,5 ml vials
    Yellow
    130×130×45 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 35,40 €
    35,40 €
    1 * 1 items
       35,40 €
  • 548-1447A
    Box with 81 cavities for 1,5 ml vials
    Box with 81 cavities for 1,5 ml vials
    Green
    130×130×45 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 35,40 €
    35,40 €
    1 * 1 items
       35,40 €
  • 548-1448A
    Box with 81 cavities for 1,5 ml vials
    Box with 81 cavities for 1,5 ml vials
    Transparent
    130×130×45 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 35,40 €
    35,40 €
    1 * 1 items
       35,40 €
16 Cavities
Product Information
  • Colour
    W×D×H
    Availability
    Price
  • 548-1995A
    Box with 16 cavities for 1,5 ml vials/2 ml shell vials
    Box with 16 cavities for 1,5 ml vials/2 ml shell vials
    Blue
    67×67×45 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 27,60 €
    27,60 €
    1 * 1 items
       27,60 €
  • 548-1996A
    Box with 16 cavities for 1,5 ml vials/2 ml shell vials
    Box with 16 cavities for 1,5 ml vials/2 ml shell vials
    Orange
    67×67×45 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 27,60 €
    27,60 €
    1 * 1 items
       27,60 €
  • 548-1997A
    Box with 16 cavities for 1,5 ml vials/2 ml shell vials
    Box with 16 cavities for 1,5 ml vials/2 ml shell vials
    Pink
    67×67×45 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 27,60 €
    27,60 €
    1 * 1 items
       27,60 €
  • 548-1998A
    Box with 16 cavities for 1,5 ml vials/2 ml shell vials
    Box with 16 cavities for 1,5 ml vials/2 ml shell vials
    Yellow
    67×67×45 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 27,60 €
    27,60 €
    1 * 1 items
       27,60 €
  • 548-1999A
    Box with 16 cavities for 1,5 ml vials/2 ml shell vials
    Box with 16 cavities for 1,5 ml vials/2 ml shell vials
    Green
    67×67×45 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 27,60 €
    27,60 €
    1 * 1 items
       27,60 €
  • 548-2000A
    Box with 16 cavities for 1,5 ml vials/2 ml shell vials
    Box with 16 cavities for 1,5 ml vials/2 ml shell vials
    Transparent
    67×67×45 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 27,60 €
    27,60 €
    1 * 1 items
       27,60 €
Other Boxes
Product Information
  • Colour
    W×D×H
    Availability
    Price
  • 548-1511A
    Box with 25 cavities for 5/10/20 ml headspace vials
    Box with 25 cavities for 5/10/20 ml headspace vials
    Blue
    133×133×102 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 52,40 €
    52,40 €
    1 * 1 items
       52,40 €
  • 548-1512A
    Box with 16 cavities for 20 ml EPA vials
    Box with 16 cavities for 20 ml EPA vials
    Violet
    133×133×70-80 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 52,40 €
    52,40 €
    1 * 1 items
       52,40 €
  • 548-1513A
    Box with 10 cavities for 30/40 ml EPA vials
    Box with 10 cavities for 30/40 ml EPA vials
    Violet
    133×133×105 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 60,70 €
    60,70 €
    1 * 1 items
       60,70 €