Order Entry
Ireland
ContactUsLinkComponent
Fluoribacter bozemanae NCTC 11368 Medium Count LENTICULE® Discs, Supelco®
Fluoribacter bozemanae NCTC 11368 Medium Count LENTICULE® Discs, Supelco®
Catalog # CRM11368M-10EA
Supplier:  Merck
CAS Number:  
Fluoribacter bozemanae NCTC 11368 Medium Count LENTICULE® Discs, Supelco®
Catalog # CRM11368M-10EA
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10
  • Description:
    Fluoribacter bozemanae NCTC 11368 Medium count LENTICULE® discs 500-50000 CFU
  • M-Clarity quality level:
    MQ300
  • Storage temperature:
    –20 °C
  • Product type:
    Hydrosoluble discs

Specifications

About this item

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.
Caution: Lenticule is a registered trademark of United Kingdom Secretary of State for Health.