Order Entry
Ireland
ContactUsLinkComponent
Citrobacter freundii WDCM 00006 Vitroids™, Certified Reference Materials, Suitable for Microbiology, Supelco®
Citrobacter freundii WDCM 00006 Vitroids™, Certified Reference Materials, Suitable for Microbiology, Supelco®
Catalog # VT000066-10EA
Supplier:  Merck
CAS Number:  
Citrobacter freundii WDCM 00006 Vitroids™, Certified Reference Materials, Suitable for Microbiology, Supelco®
Catalog # VT000066-10EA
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10
  • Description:
    Citrobacter freundii WDCM 00006 Vitroids™, 1000 - 10000 CFU mean value range
  • Seq:
    14
  • M-Clarity quality level:
    MQ300
  • Storage temperature:
    –20 °C
  • Product type:
    Hydrosoluble discs

Specifications

About this item

Derived from CECT® strains.

Certifications: These certified reference materials (CRMs) are produced and certified in accordance with ISO 17034 and ISO/IEC 17025.