Order Entry
Ireland
ContactUsLinkComponent
Staphylococcus aureus WDCM 00035 Vitroids™, Certified Reference Material, Suitable for Microbiology
Staphylococcus aureus WDCM 00035 Vitroids™, Certified Reference Material, Suitable for Microbiology
Catalog # VT000357-10EA
Supplier:  Merck
CAS Number:  
Staphylococcus aureus WDCM 00035 Vitroids™, Certified Reference Material, Suitable for Microbiology
Catalog # VT000357-10EA
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10
  • Description:
    Staphylococcus aureus WDCM 00035 Vitroids™, 50’000-150’000 CFU mean value range
  • Storage conditions:
    −20 °C
  • M-Clarity quality level:
    MQ300

Specifications

About this item