0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Ireland
ContactUsLinkComponent
Tris(perfluorobutyl)amine
Tris(perfluorobutyl)amine sample (PFTBA) for gas chromatography, for GC-MS
Tris(perfluorobutyl)amine sample (PFTBA) for gas chromatography, for GC-MS
HEWL8500-0656
:
Agilent
:
Tris(perfluorobutyl)amine sample (PFTBA) for gas chromatography, for GC-MS
HEWL8500-0656
:
Agilent
:
Pk:
10 g
Seq:
3236
:
:
671.1 g/mol
:
GHS07
:
311-89-7
:
C₁₂F₂₇N
:
MFCD00000436