Order Entry
Ireland
ContactUsLinkComponent
VWR® Maximum Protection, Cleanroom Sleeve Protectors, Sterile
VWR® Maximum Protection, Cleanroom Sleeve Protectors, Sterile
Supplier:  VWR Collection
Certificates
undefined
VWR® Maximum Protection, Cleanroom Sleeve Protectors, Sterile
Supplier:  VWR Collection
Specifications
  • Catalog No:
  • 113-9998
  • 113-9997
  • Garment application:
  • Cleanroom
  • Cleanroom
  • Particle shedding (Helmke Drum):
  • Level 1
  • Level 1
  • Thickness:
  • 0,21 mm
  • 0,21 mm
  • Weight:
  • 56 g/m²
  • 56 g/m²