Specifications
- Description:Stealth™ 9100 Safety goggle
- Lenses:Polycarbonate
- Pk:1
Specifications
About this item
A soft dual injection goggle providing a comfortable universal fit with a high level of protection from splash, dust, and impact.
- Anti-mist and anti-scratch lens
- TPV vented inner frame
- Grade 1 optics lens
- Ultra-soft housing
- Wide elastic headband with easy quick release clipping buckle
Large, injection moulded spherical wraparound optically perfect lens provides a clear field of vision. With anti-mist, anti-scratch, and Grade 1 optics. Clear lens also blocks 99.9% of ultraviolet radiation. Ultra-soft housing and dual injection frame provides a soft comfortable fit that moulds to fit almost any face shape. Wide elastic headband with easy quick release clipping buckle ensures a comfortable and suitable fit for most head sizes.
Comes complete with black, soft, elasticated military style protection sleeve to prevent lens becoming scratched when not in use. Sleeve can remain attached or removed as required.