ESD easy safety sandals, Velcro® fastening, S1P, Impulse Lady XXTL Low
Supplier: ELTEN
Certificates
About this item
Ladies' safety sandal is suitable for work in ESD sensitive or electrostatically protected areas (EPA).
- Padded tongue
- PU/PU sole WELLMAXX TRAINERS LADY
- Steel toe cap
- Leather-free
- Velcro® closure
Microfibre construction with fabric lining for natural ventilation. Full-length inlay ESD sole, PRO Lady Y. Non-metallic, penetration-resistant midsole. Sole core made of Infinergy® by BASF.
Certifications: EN ISO 20345 S1P SRC, form A.
Specifications
- Catalog No:
- ELTN741301/38
- ELTN741301/39
- ELTN741301/40
- ELTN741301/41
- ELTN741301/42
- ELTN741301/35
- ELTN741301/36
- ELTN741301/37
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001