ESD safety shoes, lace-up, Marty XXSports Low
Supplier: ELTEN
Certificates
About this item
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA).
- Padded tongue
- TPU/PU sole WELLMAXX SPORTS
- Steel toe cap
- Leather-free
Hydrophobised CORDURA® textile material construction with fabric lining for natural ventilation. Full-length inlay SPORTIVE ESD sole. Non-metallic, penetration-resistant midsole. Sole core made of Infinergy® by BASF.
Colour: Black/white/blue
Certifications: EN ISO 20345 S3 SRA, Form A.
Specifications
- Catalog No:
- ELTN728181/41
- ELTN728181/40
- ELTN728181/47
- ELTN728181/48
- ELTN728181/44
- ELTN728181/45
- ELTN728181/42
- ELTN728181/43
- ELTN728181/46
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001