Order Entry
Ireland
ContactUsLinkComponent
Human mesenchymal stem cells- Wharton's Jelly (hMSC-WJ), CELLvo™
Human mesenchymal stem cells- Wharton's Jelly (hMSC-WJ), CELLvo™
Supplier:  STEMBIOSYS
Certificates
undefined
Human mesenchymal stem cells- Wharton's Jelly (hMSC-WJ), CELLvo™
Supplier:  STEMBIOSYS

Specifications

  • Storage conditions:
    CELLvo™ XF Matrix should be stored at 2...8 °C and avoid extended exposure to light
  • Storage buffer:
    CryoStor®CS5
  • Tested applications:
    Multilineage differentiation, flow cytometry and viability

Specifications

Frequently Bought Together