Order Entry
Ireland
ContactUsLinkComponent
 
Pyridine-[D₅] (99,5% D) for NMR spectroscopy
  87095.0007
 :  
undefined
Pyridine-[D₅] (99,5% D) for NMR spectroscopy
  87095.0007
 :  

 

  • Pack type:
    Glass ampoule
  • Pk:
    1 SET
  • Seq:
    2065
  • Environmentally Preferable:
  • Pk Info:
    10× 0,75 ml

 

 

  • Purity & Structure:
    conforms
  • Isotopic enrichment:
    min. 99,5% D
  • Water content:
    max. 0,05%

 

 

 : Pyridine-[²H₅]
 :
 :84.06 g/mol
 :1282
 :GHS02
 :GHS07
 :Ambient
 :7291-22-7
 :C₅D₅N
 :MFCD00044639
 :114.4
 :-41
 :20
 :3 II