Water with 0,05% (v/v) trifluoroacetic acid, HiPerSolv CHROMANORM® for LC-MS
Catalog # 88379.320
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Nominal concentration (v/v):0.045 - 0.055%
- Evaporation residue:max. 0.0001%
- Ca (Calcium):max. 0.2 ppm
- K (Potassium):max. 0.2 ppm
- Mg (Magnesium):max. 0.2 ppm
- Na (Sodium):max. 0.5 ppm
- Transmittance (210 nm):min. 20%
- Transmittance (220 nm):min. 50%
- Transmittance (230 nm):min. 80%
- Transmittance (240 nm):min. 95%
- Transmittance (250 nm):min. 99%
- Suitable for LC-MS:passes test
Specification Test Results
About this item
:
Storage Temperature:Ambient
:76-05-1