


Chlorobenzene ≥99.5%, AR®, Macron Fine Chemicals™
Catalog # 6788-25
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Assay (GC):≥99,5%
- Water (KF):≤0,03%
- Titrable Acid (meq/g) :≤0,0005
- Residue after Evaporation:≤0,003%
Specification Test Results
About this item
:
:112.56 g/mol
Melting Pt:–45 °C
Density:1,11 g/cm³ (20 °C)
Flash Pt:28 °C
Storage Temperature:Ambient
EINECS:203-628-5
UN:1134
ADR:3,III
:GHS02
:GHS07
:GHS09
CAS Number:108-90-7
Formula:C₆H₅Cl
MDL Number:MFCD00000530