Potassium cyanide ≥96.0%, AnalaR NORMAPUR® ACS analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:151-50-8
:
Formula:KCN
MDL Number:MFCD00011397
:65.12 g/mol
Storage Temperature:Ambient
EINECS:205-792-3
UN:1680
ADR:6.1,I
:GHS05
:GHS06
:GHS08
:GHS09
Specification Test Results
- Appearance:White crystalline powder or crystals
- Assay:min. 96.0%
- Chloride (Cl):max. 0.5%
- Sulfate (SO4):max. 0.04%
- Sulfide (S):max. 0.003%
- Phosphate (PO4):max. 0.005%
- Thiocyanate (SCN):passes test (limit about 0.02%)
- Iron (Fe):max. 0.03%
- Lead (Pb):max. 0.0002%
- Sodium (Na):max. 0.5%
Specification Test Results
Frequently Bought Together