Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Electrodes
pH Electrodes
Accessories for pH electrodes, SteamLine
Accessories for pH electrodes, SteamLine
Accessories for pH electrodes, SteamLine
Catalog # 662-6164
Supplier:
SI Analytics
Expand image
Accessories for pH electrodes, SteamLine
Catalog # 662-6164
Supplier:
SI Analytics
Specifications
Description:
9902/05 cable for coaxial plug head (SL80, SL89), 5 m
Pk:
1
Specifications
About this item
Product Details
Documentation
Product Details
Documentation