Order Entry
Ireland
ContactUsLinkComponent
Water, Cell Culture Grade pyrogen-free
Water, Cell Culture Grade pyrogen-free
Supplier:  VWR Chemicals
Certificates
undefined
Water, Cell Culture Grade pyrogen-free
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Color:
    colourless, clear solution
  • Endotoxins:
    < 0.005 EU/ml
  • Sterility test:
    Bacteria in aerobic and anaerobic conditions
  • Shelf life:
    48 months

Specification Test Results

Frequently Bought Together