C18 EWP plates, Bond Elut
Supplier: VARIAN
Certificates
About this item
Bond Elut C18 EWP uses a special silica with wide pores. This is endcapped.
- No exclusion of large molecules
- Good for desalting proteins
- Successful separation of proteins, peptides or nucleotides
This is based upon standard particles size silica but with 500 Å pores to allow more efficient extraction of large molecules (>15000 MW), which are typically excluded from standard porosity silica phases.
Frequently Bought Together